Human recombinant APRIL (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human APRIL / TNFSF13 protein, His,Flag Tag, active trimer (MALS verified).
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:active trimer (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Loaded Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) on HIS1K Biosensor, can bind Human BCMA, Fc Tag (Cat. No. BC7-H5254) with an affinity constant of 1,88 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) on HIS1K Biosensor, can bind Human TACI/TNFRSF13B, Fc Tag (Cat. No. TAI-H5256) with an affinity constant of 75 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:O75888-1
- Protein synonyms:APRIL|TNFSF13|TALL2|TRDL-1|ZTNF2|TALL-2|CD256
- Protein/peptide name:APRIL
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:50,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together