Order Entry
Belgium
ContactUsLinkComponent
 
Rabbit recombinant FcRn (FCGRT & B2M) (from HEK293 cells)
Catalog # ACRBFCM-R52W5-50UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Rabbit recombinant FcRn (FCGRT & B2M) (from HEK293 cells)
Catalog # ACRBFCM-R52W5-50UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rabbit
  • Tag sequence:
    His Tag&Tag Free (SPR verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Rabbit FCGRT and B2M Heterodimer protein, His Tag and Tag Free (Cat. No. FCM-R52W5) captured on CM5 Chip via anti-His antibody can bind Herceptin® with an affinity constant of 2,12 μM as determined in SPR assay (Biacore T200) (QC tested).
  • Protein synonyms:
    FcRn|FCGRT & B2M
  • Protein/peptide name:
    FcRn (FCGRT & B2M)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    32,1 kDa (FCGRT) and 11,7 kDa (B2M)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Rabbit FcRn / FCGRT&B2M Heterodimer protein, His Tag&Tag Free (SPR verified).