Mouse recombinant TRAIL R2 (from HEK293 cells)
Catalog # ACRBTR2-M52H5-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human TNFSF10 at 2 μg/ml (100 μl/well) can bind Mouse TRAIL R2, His Tag (Cat. No. TR2-M52H5) with a linear range of 0,039 - 0,625 μg/ml (QC tested).
- Gene ID:Q9QZM4-1
- Protein synonyms:TRAILR2|TRAIL-R2|CD262|TRICK2|KILLER|TRICKB|ZTNFR9|TNFRSF10B|DR5
- Protein/peptide name:TRAIL R2
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:16,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Mouse TRAIL R2 / DR5 / TNFRSF10B protein, His Tag.