Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant Glypican 3 (from HEK293)
Mouse recombinant Glypican 3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant Glypican 3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human FGF basic, Tag Free at 2 μg/ml (100 µl/well) can bind Mouse Glypican 3, Fc Tag with a linear range of 1-10 ng/ml (QC tested).
  • Gene ID:
    Q8CFZ4-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
  • Protein/peptide name:
    Glypican 3
  • Purity:
    >90%
  • Molecular weight:
    38,0 kDa and 49,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together