Mouse recombinant GITR ligand (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Mouse GITR, His Tag at 5 μg/ml (100 µl/well) can bind Mouse GITR Ligand, Fc Tag with a linear range of 5-156 ng/ml (QC tested).
- Gene ID:Q7TS55(T157N)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:hGITRL|TL6|AITRL|TNFSF18|GITR Ligand
- Protein/peptide name:GITR Ligand
- Purity:>95%
- Molecular weight:41,4 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together