Mouse recombinant CD40 ligand (from HEK293)
Fournisseur: ACRO Biosystems
Certificats
À propos de cet article
Spécifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Mouse CD40, His Tag at 5 μg/ml (100 µl/well) can bind Mouse CD40 Ligand, Fc Tag with a linear range of 1-16 ng/ml (QC tested).
- Gene ID:P27548-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Protein/peptide name:CD40 Ligand
- Purity:>95%
- Molecular weight:42,9 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Spécifications
Frequently Bought Together