Order Entry
Belgium
ContactUsLinkComponent
Human recombinant TIM-3 (from HEK293)
Human recombinant TIM-3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant TIM-3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method.
  • Biological activity:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human TIM-3, Mouse IgG2a Fc Tag, low endotoxin at 0,2 μg/ml (100 µl/well) can bind Anti-TIM3 MAb, Human IgG4 with a linear range of 0,2-2 ng/ml (QC tested).
  • Gene ID:
    Q8TDQ0-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TIMD3|FLJ14428|KIM3|TIM3|HAVCR2
  • Protein/peptide name:
    TIM-3
  • Purity:
    >95%
  • Molecular weight:
    46,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together