Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Siglec-3 (from HEK293)
Human recombinant Siglec-3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant Siglec-3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,01 EU per μg by the LAL method.
  • Biological activity:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin at 5 μg/ml (100 µl/well) can bind Anti-CD33, Human IgG1 with a linear range of 0,078-1,25 μg/ml (QC tested).; Loaded Siglec-3 MAb (Mouse IgG1) on AMC Biosensor, can bind Human Siglec-3, Llama IgG2b Fc Tag, low endotoxin with an affinity constant of 0,365 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P20138-1(R69G)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    gp67|SIGLEC3|CD33
  • Protein/peptide name:
    Siglec-3
  • Purity:
    >95%
  • Molecular weight:
    54,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together