Order Entry
Belgium
ContactUsLinkComponent
Human recombinant NKG2C & CD94 (from HEK293 cells)
Human recombinant NKG2C & CD94 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant NKG2C & CD94 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P26717-1 (NKG2C) & Q13241-1 (CD94)
  • Protein/peptide name:
    NKG2C & CD94
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    57,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together