Human recombinant mucin-1 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P15941-1
- Protein synonyms:Mucin 1|Episialin|CA 15-3|PEMT|PUM|MUC-1/X|CD227|MUC-1/SEC|KL-6|EMA|MUC1|MAM6|PEM|H23AG|MUC1/ZD
- Protein/peptide name:Mucin-1
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:60,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together