Order Entry
Belgium
ContactUsLinkComponent
Human recombinant LILRB4 (from HEK293 cells)
Human recombinant LILRB4 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant LILRB4 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (SPR verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Human ANGPTL7, His Tag Immobilised on CM5 Chip can bind Human LILRB4, His Tag (SPR verified) with an affinity constant of 4,67 μm as determined in a SPR assay (Biacore T200) (QC tested).
  • Protein synonyms:
    CD85K|HM18|ILT3|LIR5|LILRB4
  • Protein/peptide name:
    LILRB4
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    28,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together