Order Entry
Belgium
ContactUsLinkComponent
Human recombinant integrin alpha V beta 1 (from HEK293 cells)
Human recombinant integrin alpha V beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant integrin alpha V beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Tag Free&Tag Free
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Recombinant Human Fibronectin at 2 μg/ml (100 μl/well) can bind Human ITGAV&ITGB1 Heterodimer Protein, Tag Free&Tag Free with a linear range of 0,002 to 0,313 μg/ml (QC tested).
  • Protein synonyms:
    Integrin alpha V beta 1|ITGAV&ITGB1
  • Protein/peptide name:
    Integrin alpha V beta 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    112,3 kDa (ITGAV) and 83,7 kDa (ITGB1)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8,0. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together