Mouse recombinant Fc gamma RIIB/ CD32b (from HEK293 cells)
Catalog # ACRBCDB-M52H7-50UG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pk:50 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag (SPR & BLI verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Immobilised Mouse CD32b, His Tag (SPR & BLI verified) (ACRBCDB-M52H7) on NTA-Series S sensor chip, can bind OKT3 with an affinity constant of 0.86 μM as determined in a SPR assay (Biacore T200) (QC tested). Immobilised OKT3 on Protein G-Series S sensor chip, can bind Mouse CD32b, His Tag (SPR & BLI verified) (ACRBCDB-M52H7) with an affinity constant of 0,55 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Mouse CD32b, His Tag (SPR & BLI verified) (ACRBCDB-M52H7) on NTA Biosensor, can bind OKT3 with an affinity constant of 1,2 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Mouse CD32b, His Tag (SPR & BLI verified) (ACRBCDB-M52H7) on NTA Biosensor, can bind Herceptin with an affinity constant of 2,1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded OKT3 on Protein G Biosensor, can bind Mouse CD32b, His Tag (SPR & BLI verified) (Cat. No. CDB-M52H7) with an affinity constant of 1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Herceptin on Protein G Biosensor, can bind Mouse CD32b, His Tag (SPR & BLI verified) (Cat. No. CDB-M52H7) with an affinity constant of 1,2 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P08101
- Protein synonyms:CD32|IGFR2|CD32b|FCG2|CDw32|FCGR2B
- Protein/peptide name:Fc gamma RIIB/ CD32b
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:22,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Mouse Fc gamma RIIB / CD32b Protein, His Tag (SPR & BLI verified).