Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD36 (from HEK293 cells), biotin
Human recombinant CD36 (from HEK293 cells), biotin
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant CD36 (from HEK293 cells), biotin
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Protein synonyms:
    GP4|GP3B|FAT|GPIV|gpIV|SCARB3|CHDS7|glycoprotein IIIb|CD36|glycoprotein IV|BDPLT10|gpIIIb|PASIV|Platelet Glycoprotein 4
  • Protein/peptide name:
    CD36
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    50,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together