Saisie de commande
Belgium
ContactUsLinkComponent
Human recombinant LILRB3 (from HEK293)
Human recombinant LILRB3 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant LILRB3 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugaison:
    Unconjugated
  • Type de protéine/peptide:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Conditions de stockage:
    –20 °C
  • Teneur en endotoxines:
    Less than 1,0 EU per μg by the LAL method.
  • Activité biologique:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Sans vecteur:
    Y
  • Exempt de protéase:
    Y
  • Exempt de substance animale:
    Y
  • Synonyme de protéine:
    ILT5|LILRB3|CD85a
  • Nom de la protéine/du peptide:
    LILRB3
  • Purity:
    >92%
  • Molecular weight:
    48 kDa
  • Niveau d'endotoxine:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Absence de nucléases:
    Y
  • Température de transport:
    Powder,RT

Specifications

Frequently Bought Together