Order Entry
Belgium
ContactUsLinkComponent
Human recombinant latent TGF-beta 1 (from HEK293)
Human recombinant latent TGF-beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant latent TGF-beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised human latent TGFB1, His Tag (Cat. No. ACRBTG1-H524x) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB6 heterodimer protein, His,Avitag and Tag Free (Cat. No. ACRBIT6-H82E4) with a linear range of 1 - 20 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised human latent TGFB1, His Tag (Cat. No. ACRBTG1-H524x) at 5 μg/ml (100 μl/well) can bind human LRRC32, Fc Tag (Cat. No. ACRBLR2-H5256) with a linear range of 0,039 - 0,625 μg/ml (routinely tested). Human latent TGFB1, His Tag (Cat. No. ACRBTG1-H524x) inhibits the IL-4-dependent proliferation of TF-1 cells. The ED50 for this effect is 0,293 - 0,406 ng/ml (routinely tested).
  • Gene ID:
    P01137-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CED|TGF-beta-1|TGFB|DPD1|Latent TGF-beta 1|Latent TGFB1|LAP|TGFB1
  • Protein/peptide name:
    Latent TGF-beta 1
  • Purity:
    >95%
  • Molecular weight:
    42,6 kDa (monomer)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together