Order Entry
Belgium
ContactUsLinkComponent
Human recombinant LAP (TGF-beta 1) (from HEK293)
Human recombinant LAP (TGF-beta 1) (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant LAP (TGF-beta 1) (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 . Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. Immobilised Human LAP (TGF-beta 1), His Tag (Cat. No. ACRBLAP-H5245) at 1 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB6 heterodimer protein, His, Avitag and tag free (Cat. No. ACRBIT6-H82E4) with a linear range of 2 - 31 ng/ml (QC tested).
  • Gene ID:
    P01137-1 (C33S)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CED|TGF-beta-1|LAP (TGF-beta 1)|LAP (TGFB1)|TGFB|DPD1|LAP|TGFB1
  • Protein/peptide name:
    LAP (TGF-beta 1)
  • Purity:
    >95%
  • Molecular weight:
    40 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 . Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together