Order Entry
Belgium
ContactUsLinkComponent
Human recombinant integrin alpha V beta 6 (from HEK293)
Human recombinant integrin alpha V beta 6 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant integrin alpha V beta 6 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human TGFB1, His Tag (Cat. No. ACBRTG1-H524x) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB6 heterodimer protein, His, Avitag and tag free (Cat. No. ACBRIT6-H82E4) with a linear range of 2 - 10 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised fibronectin at 10 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB6 heterodimer protein, His,Avitag and tag free (Cat. No. ACBRIT6-H82E4) with a linear range of 0,08 - 1,25 μg/ml (routinely tested).
  • Gene ID:
    NP_002201 (ITGAV) and P18564-1 (ITGB6)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    ITGAV and ITGB6|Integrin alpha V beta 6
  • Protein/peptide name:
    Integrin alpha V beta 6
  • Purity:
    >90%
  • Molecular weight:
    114,7 kDa (ITGAV) and 79,6 kDa (ITGB6)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together