Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant IL-17F (from HEK293)
  ACRBILF-H4240-20UG
 :  
Human recombinant IL-17F (from HEK293)
  ACRBILF-H4240-20UG
 :  

 

  • Pk:
    20 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; loaded human IL-17 RA, Fc Tag (Cat. No.ACRBILA-H5257) on protein A biosensor, can bind Human IL-17F (H161R), His Tag (Cat. No. ACRBILF-H4240) with an affinity constant of 299 nM as determined in BLI assay (ForteBio Octet Red96e) (routinely tested).
  • Gene ID:
    Q96PD4-1 (H161R)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IL24|IL-17F|ML-1|Interleukin-17F
  • Protein/peptide name:
    IL-17F
  • Purity:
    >95%
  • Molecular weight:
    16,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT