Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-12B (from HEK293)
Human recombinant IL-12B (from HEK293)
Catalog # ACRBNK2-H52H7-20UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant IL-12B (from HEK293)
Catalog # ACRBNK2-H52H7-20UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilized Anti IL23A and IL12B MAb at 2 μg/ml (100 μl/well) can bind human IL-12B, His Tag (Cat. No. ACRBNK2-H52H7) with a linear range of 0,4 - 13 ng/ml (QC tested). Loaded Human IL-12 R beta 1, Fc Tag (Cat. No. ACRBILB-H5255) on protein A biosensor, can bind human IL-12B, His Tag (Cat. No. ACRBNK2-H52H7) with an affinity constant of 3,78 nM as determined in BLI assay (ForteBio Octet Red96e) (routinely tested). Loaded Anti-IL23A and IL12B MAb (P40 domain) on protein A biosensor, can bind human IL-12B, His Tag (Cat. No. ACRBNK2-H52H7) with an affinity constant of 0,279 nM as determined in BLI assay (ForteBio Octet Red96e) (routinely tested).
  • Gene ID:
    P29460-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    NKSF|CLMF|CLMF2|NKSF2|p40|IL12B
  • Protein/peptide name:
    IL-12B
  • Purity:
    >95%
  • Molecular weight:
    36,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item