Order Entry
Belgium
ContactUsLinkComponent
Human recombinant transferrin R (from HEK293 cells), Biotin
Human recombinant transferrin R (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant transferrin R (from HEK293 cells), Biotin
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human Transferrin, His Tag (ACRBTRN-H4229) at 2 μg/ml (100 μl/well) can bind Biotinylated Human Transferrin R, His Tag (ACRBTFR-H8243) with a linear range of 5 - 156 ng/ml (QC tested).
  • Protease-free:
    Y
  • Protein synonyms:
    TfR1|T9|Trfr|CD71|sTfR|p90|TFRC|TFR
  • UniProtKB:
    P02786
  • Protein/peptide name:
    Transferrin R
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    76 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together