Order Entry
Belgium
ContactUsLinkComponent
Cynomolgus recombinant Her2 (from HEK293)
Cynomolgus recombinant Her2 (from HEK293)
Catalog # ACRBHE2-C52HB-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Cynomolgus recombinant Her2 (from HEK293)
Catalog # ACRBHE2-C52HB-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pack type:
    1 mg
  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 02,2 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Loaded Hercecptin on AHC Biosensor, can bind Cynomolgus Her2, His Tag with an affinity constant of 4,86 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Protein/peptide name:
    Her2
  • Purity:
    >95%
  • Molecular weight:
    71,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item