Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant FAP (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
 
Human recombinant FAP (from HEK293 cells), FITC (Fluorescein Isothiocyanate)

 

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    2e5 of Anti-FAP CAR-293 cells were stained with 100 μL of 1 μg/ml FITC-Labeled Human FAP, Fc Tag (Cat. No. FAP-HF263) and negative control protein respectively, FITC signal was used to evaluate the binding activity (QC tested).
  • Gene ID:
    Q12884-1
  • Protein synonyms:
    FAP|APCE|SIMP|Seprase|FAPalpha
  • Protein/peptide name:
    FAP
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    111,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together