Human recombinant Fc epsilon RI alpha (from HEK293 cells)
Human Fc epsilon RI alpha Protein, Fc Tag (BLI verified), ACROBiosystems.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (BLI verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Loaded Human Fc epsilon RI alpha, Fc Tag (BLI verified) (Cat. No. FCA-H5259) on AHC Biosensor, can bind Immunoglobulin E, Human Plasma with an affinity constant of 0.188 nM as determined in BLI assay (ForteBio Octet Red96e) (QC Tested).
- Gene ID:P12319-1
- Protein synonyms:FCE1A|FcERI|FCER1A
- Protein/peptide name:Fc epsilon RI alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:47,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Frequently Bought Together