Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant frizzled-10 (from HEK293 cells)
 
Human recombinant frizzled-10 (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    Q9ULW2-1
  • Protein synonyms:
    FzE7|FZD10|Fz-10|CD350|hFz10
  • Protein/peptide name:
    Frizzled-10
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    18,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together