Order Entry
Belgium
ContactUsLinkComponent
Cynomolgus monkey recombinant CD73 (from HEK293 cells)
Cynomolgus monkey recombinant CD73 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Cynomolgus monkey recombinant CD73 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Tag sequence:
    His Tag (active enzyme)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Protein synonyms:
    5'-Nucleotidase|5'-NT|NTE|NT5|NT5E|CD73
  • Protein/peptide name:
    CD73
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    60,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together