Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-1 RAcP (from HEK293 cells)
Human recombinant IL-1 RAcP (from HEK293 cells)
Catalog # ACRBILP-H5225-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant IL-1 RAcP (from HEK293 cells)
Catalog # ACRBILP-H5225-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pack type:
    1 mg
  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein function:
    Bio-Markers & CD Antigens
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IL1RAP|IL1R3|IL-1RAcP|C3orf13
  • UniProtKB:
    Q9NPH3
  • Protein/peptide name:
    IL-1 RAcP
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    40 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    Powder,RT

Specifications

About this item