Order Entry
Belgium
ContactUsLinkComponent
Human recombinant siglec-3 (from HEK293 cells), FITC (fluorescein isothiocyanate)
Human recombinant siglec-3 (from HEK293 cells), FITC (fluorescein isothiocyanate)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant siglec-3 (from HEK293 cells), FITC (fluorescein isothiocyanate)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 ℃, aviod light
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    P20138-1 (R69G)
  • Protein synonyms:
    gp67|SIGLEC3|CD33
  • Protein/peptide name:
    Siglec-3
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    27,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together