Human recombinant siglec-15 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Siglec-15 / CD33L3 Protein, His Tag.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 5 μg/ml (100 μl/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0,3 - 2 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-15, His Tag (Cat. No. SG5-H52H3) at 10 μg/ml (100 μl/well) on Nickel Coated plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0.078-5 μg/ml (Routinely tested). Loaded Mouse Human chimeric MAb (5G12, Human IgG1) on AHC Biosensor, can bind Human Siglec-15, His Tag (Cat. No. SG5-H52H3) with an affinity constant of 0,52 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q6ZMC9-1
- Protein synonyms:Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
- Protein/peptide name:Siglec-15
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:28,4 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH 5,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together