Human recombinant siglec-15 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human Siglec-15 / CD33L3 Protein, Fc Tag.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-15, Fc Tag (Cat. No. SG5-H5253) at 5 μg/ml (100 μl/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0,4 - 6 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human Siglec-15, Fc Tag (Cat. No. SG5-H5253) at 10 μg/ml (100 μl/well) on Diamond Protein A Protein, His Tag precoated (0,5 μg/well) plate, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0,078 - 1,25 μg/ml (Routinely tested). Immobilised Neu5Ac(a2-6)GalNAc-PAA-biotin on SA Chip can bind Human Siglec-15, Fc Tag (Cat. No. SG5-H5253) with an affinity constant of 4,49 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Human Siglec-15, Fc Tag (Cat. No. SG5-H5253) on ProteinA Biosensor, can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with an affinity constant of 0,78 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Anti-Siglec-15 MAb (Mouse IgG) on AMC Biosensor, can bind Human Siglec-15, Fc Tag (Cat. No.SG5-H5253) with an affinity constant of 59,7 pM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Human Siglec-15, Fc Tag (Cat. No. SG5-H5253) inhibits Anti-CD3-induced proliferation of PBMC. The ED50 for this effect is 0,46 - 0,54 μg/ml (Routinely tested).
- Gene ID:Q6ZMC9-1
- Protein synonyms:Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
- Protein/peptide name:Siglec-15
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:53,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together