Human recombinant siglec-15 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Siglec-15 / CD33L3 Protein, Mouse IgG2a Fc Tag.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Mouse IgG2a Fc Tag
- Storage conditions:–20 °C
- Gene ID:Q6ZMC9-1
- Protein synonyms:Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
- Protein/peptide name:Siglec-15
- Molecular weight:53,4 kDa
- Endotoxin level:Low
- Formulation:Please contact us for detailed information.
- Shipping temperature:RT
Specifications
Frequently Bought Together