Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant siglec-15 (from HEK293 cells), Biotin
 
Human recombinant siglec-15 (from HEK293 cells), Biotin

 

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Mouse Human chimeric MAb (5G12, Human IgG1) at 5 μg/ml (100 μl/well) can bind Biotinylated Human Siglec-15, His,Avitag (recommended for biopanning) (Cat. No. SG5-H82E9) with a linear range of 0,2 - 3 ng/ml (QC tested).
  • Gene ID:
    Q6ZMC9-1
  • Protein synonyms:
    Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
  • Protein/peptide name:
    Siglec-15
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    30,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 25 mM MES,150 mM NaCl, pH 5,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together