Human recombinant siglec-1 (from HEK293 cells)
Human Siglec-1 / CD169 Protein, His Tag.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:Q9BZZ2-1
- Protein synonyms:SIGLEC1|Siglec-1|SN|CD169
- Protein/peptide name:Siglec-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:175,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in MES, NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together