Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ROR1 (from HEK293 cells)
Human recombinant ROR1 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant ROR1 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Mouse IgG2a Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method
  • Gene ID:
    Q01973-1
  • Protein synonyms:
    NTRKR1|ROR1
  • Protein/peptide name:
    ROR1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    68,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together