Order Entry
Belgium
ContactUsLinkComponent
 
Human recombinant PDGF R alpha (from HEK293 cells)
 
Human recombinant PDGF R alpha (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Human PDGF-BB, Tag Free (Cat. No. PDB-H4112) at 2 μg/ml (100 μl/well) can bind Human PDGF R alpha, His Tag (Cat. No. PDA-H52H7) with a linear range of 0,02 - 0,625 μg/ml (QC tested).
  • Gene ID:
    P16234-1
  • Protein synonyms:
    PDGFRA|CD140A|RHEPDGFRA|PDGFR2|MGC74795
  • Protein/peptide name:
    PDGF R alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    58,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together