Cynomolgus monkey recombinant CD24 (from HEK293 cells)
Leverancier: ACROBIOSYSTEMS
Certificaten
Over dit artikel
Cynomolgus CD24 Protein, Fc Tag.
Specificaties
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:I7GKK1-1
- Protein synonyms:CD24A|CD24
- Protein/peptide name:CD24
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:29,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specificaties
Frequently Bought Together