Human recombinant EGF R (from HEK293 cells)
Catalog # ACRBEGR-H5222-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein function:Bio-Markers & CD Antigens
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cetuximab at 2 μg/ml (100 μl/well) can bind Human EGF R, His Tag with a linear range of 0,8-6 ng/ml (QC tested).; Erbitux (Cetuximab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human EGF R, His Tag with an affinity constant of 1,3 nM as determined in a SPR assay (Biacore T200) (Routinely tested)
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:PIG61|HER1|mENA|EGFR|ERBB|ERBB1
- UniProtKB:P00533-1
- Protein/peptide name:EGF R
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:70,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilized from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:Powder,RT