Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Fc gamma RIIIB / CD16b (NA1) (from HEK293)
Human recombinant Fc gamma RIIIB / CD16b (NA1) (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant Fc gamma RIIIB / CD16b (NA1) (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Biotinylated Human CD16b (NA1), His,Avitag (SPR and BLI verified) captured on Biotin CAP- Series S Sensor Chip can bind Herceptin with an affinity constant of 8,32 μM as determined in a SPR assay (Biacore T200) (QC tested). Immobilised Biotinylated Human CD16b (NA1), His,Avitag (SPR and BLI verified) on SA Chip, can bind Herceptin with an affinity constant of 3,31 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Biotinylated Human CD16b (NA1), His,Avitag (SPR and BLI verified) (ACRBCDB-H82E4-25UG) on SA Biosensor, can bind Rituximab with an affinity constant of 7,7 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    O75015-1 (S36R, S65N, N82D, I106V)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    FCGR3B|FCGR3|FCG3B|FCG3|CD16B|IGFR3|Fc gamma RIIIB / CD16b (NA1)
  • Protein/peptide name:
    Fc gamma RIIIB / CD16b (NA1)
  • Purity:
    >95%
  • Molecular weight:
    24,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together