Mouse recombinant Siglec-2 (from HEK293)
Fournisseur: ACROBIOSYSTEMS
Certificats
À propos de cet article
Spécifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:P35329-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:BL-CAM|SIGLEC-2|CD22|Siglec2|SIGLEC2|SIGLEC2FLJ22814
- Protein/peptide name:Siglec-2
- Purity:>95%
- Molecular weight:78,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Spécifications
Frequently Bought Together