Mouse recombinant Siglec-15 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Mouse Siglec-15 / CD33L3 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Mouse Siglec-15, His Tag (Cat. No. SG5-M52H7) at 5 μg/ml (100 μl/well) can bind Mouse Human chimeric MAb (5G12, Human IgG1) with a linear range of 0,3 - 2 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Mouse Siglec-15, His Tag (Cat. No. SG5-M52H7) at 10 μg/ml (100 μl/well) can bind Neu5Ac(a2-6)GalNAc-PAA-biotin with a linear range of 0,078 - 1,25 μg/ml (Routinely tested).
- Gene ID:A7E1W8-1
- Protein synonyms:Siglec-15|SIGLEC-15|Siglec15|CD33L3|CD33 antigen-like 3|sialic acid-binding Ig-like lectin 15
- Protein/peptide name:Siglec-15
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:27,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS with Arginine, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifications
Frequently Bought Together