Mouse recombinant Mesothelin (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Mouse Mesothelin / MSLN (298-600) Protein, His Tag (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:Q61468-1
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:36,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
- Shipping temperature:RT
Specifications
Frequently Bought Together