Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-2 R beta & IL-2 R gamma (from HEK293 cells)
Human recombinant IL-2 R beta & IL-2 R gamma (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant IL-2 R beta & IL-2 R gamma (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag&Fc Tag (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P14784-1 (IL-2RB) & P31785-1 (IL-2RG)
  • Protein synonyms:
    IL-2RB & IL-2RG|IL-2 R beta & IL-2 R gamma
  • Protein/peptide name:
    IL-2 R beta & IL-2 R gamma
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    51,1 kDa (IL-2RB) and 53,6 kDa (IL-2RG)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together