Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-17F (from HEK293 cells)
Human recombinant IL-17F (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant IL-17F (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Loaded Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) on Protein A Biosensor, can bind Human IL-17F, His Tag (Cat. No. ILF-H5244) with an affinity constant of 219 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human IL17RA & IL17RC Protein, Fc Tag&Fc Tag (Cat. No. ILC-H5257) on Protein A Biosensor, can bind Human IL-17F, His Tag (Cat. No. ILF-H5244) with an affinity constant of 1,04 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q96PD4-1
  • Protein synonyms:
    IL-17F|Interleukin-17F|Cytokine ML-1
  • Protein/peptide name:
    IL-17F
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    16,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together