Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-15 R alpha (from HEK293 cells)
Human recombinant IL-15 R alpha (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant IL-15 R alpha (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 μg/ml (100 μl/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2 - 78 ng/ml (QC tested). Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) inhibits IL-15-dependent proliferation of CTLL-2 cells. The ED50 for this effect is 2,77 - 4,20 ng/ml (Routinely tested).
  • Gene ID:
    Q13261-1
  • Protein synonyms:
    CD215|IL-15RA|IL-15R-alpha|IL-15 R alpha|IL15RA
  • Protein/peptide name:
    IL-15 R alpha
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    44,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together