Order Entry
Belgium
ContactUsLinkComponent
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant IL-12 R beta 1 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Human IL-12B, His Tag (Cat. No. NK2-H52H7) at 5 μg/ml (100 μl/well) can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with a linear range of 5 - 78 ng/ml (QC tested). Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) with an affinity constant of 2,06 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) on Protein A Biosensor, can bind Human IL-12B, His Tag (Cat. No. NK2-H52H7) with an affinity constant of 7,14 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human IL-12B&IL-12A Heterodimer Protein, His Tag&Flag Tag (Cat. No. IL2-H4210) on HIS1K Biosensor, can bind Human IL-12 R beta 1, Fc Tag (Cat. No. ILB-H5255) with an affinity constant of 32,3 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P42701-1
  • Protein synonyms:
    CD212|IL12RB|IL-12 R beta 1|IL12RB1|IL-12RB1|IL12R
  • Protein/peptide name:
    IL-12 R beta 1
  • Purity:
    >95% as determined by reduced SDS-PAGE.
  • Molecular weight:
    83,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together