Human recombinant Fc gamma RIIA/ CD32a (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag (BLI verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Immobilised Human CD32a (R167), His Tag (BLI verified) (Cat. No. CDA-H5221) on CM5 Chip via anti-His antibody, can bind Rituximab with an affinity constant of 3.12 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Rituximab on Protein A Biosensor, can bind Human CD32a (R167), His Tag (BLI verified) (Cat. No. CDA-H5221) with an affinity constant of 1,5 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested). Loaded Human CD32a (R167), His Tag (BLI verified) (Cat. No. CDA-H5221) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 1,4 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P12318-1 (R167)
- Protein synonyms:CD32|CD32a|IGFR2|FCGR2A1|FCG2 |FCGR2A
- Protein/peptide name:Fc gamma RIIA/ CD32a
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:21,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together