Human recombinant ErbB3 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Mouse IgG2a Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Gene ID:P21860-1
- Protein synonyms:p180-ErbB3|MDA-BF-1|HER3|MGC88033|p85-sErbB3|c-erbB3|erbB3-S|LCCS2|ERBB3|p45-sErbB3
- Protein/peptide name:ErbB3
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:95,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together