Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Coagulation factor XI (from HEK293 cells)
Human recombinant Coagulation factor XI (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant Coagulation factor XI (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (active enzyme)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P03951-1
  • Protein synonyms:
    F11|PTA|Coagulation factor XI|FXI
  • Protein/peptide name:
    Coagulation factor XI
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    69,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 25 mM HEPES, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together