Human recombinant CD8 alpha (from HEK293 cells)
ACRBCDA-H52H3-1MG
:
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P01732-1
- Protein synonyms:p32|CD8A|Leu2|MAL|CD8
- Protein/peptide name:CD8 alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:19,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT