Human recombinant CD39 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Mouse IgG2a Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Gene ID:P49961-1
- Protein synonyms:CD39|Ecto-ATPDase 1|Ecto-ATPase 1|ENTPD1|Entpd1|NTPDase 1
- Protein/peptide name:CD39
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:77,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 20 mM Tris, 500 mM Argnine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together